Learn More
Medchemexpress LLC HY-12501A 5mg Medchemexpress, ITI-214 CAS:1642303-38-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12501A5MG
Encompass_Preferred
Medchemexpress, HY-12501A 5mg ITI-214 CAS:1642303-38-5 ITI-214 is a potent, CNS-active, orally bioavailable PDE1 inhibitor (Ki of 58 pM) with excellent selectivity against other PDE family members and against a panel of enzymes, receptors, transporters and ion channels. ITI-214 inhibits recombinant full-length human PDE1A, PDE1B and PDE1C with Kis of 33 pM, 380 pM and 35 pM, respectively. ITI-214 shows efficacy in various animal models of motor and cognitive functions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1064