Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ITIC-Cl

Catalog No. 906387100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-638-7100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-638-7100MG Supplier Sigma Aldrich Supplier No. 906387100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

ITCI-CI is a low band acceptor-donor-acceptor molecule with indacenodithieno[3,2G+b]thiophene (IT) as a central donor atom and 2G+(3G+oxoG+2,3G+dihydroindenG+1G+ylidene)malononitrile (IC) as an acceptor end group. It is also a non-fullerene electron acceptor with a push-pull structure that induces an intramolecular charge transfer and extends absorption.ITIC-Cl is a novel non-fullerene acceptor material designed to use in organic photovoltaic devices.ITIC-Cl is a derivative of the ITIC molecule, which is increasingly used to replace fullerene based derivatives for organic solar cells due to significantly improved performances and stability.Compared to ITIC and its other derivatives such as ITIC-F, ITIC-Cl has a narrower bandgap and lower energy levels, and thus will be more suitable to pair with various p-type polymers.ITIC-Cl is readily soluble in most organic solvents used to fabricate organic electronic devices.

Specifications

CAS 2253663-81-7
Molecular Formula C94H78Cl4N4O2S4
Linear Formula C94H78Cl4N4O2S4
Quantity 100 mg
Synonym 3,9-bis(2-methylene-((3-(1,1-dicyanomethylene)-6,7-dichloro)-indanone))-5,5,11,11-tetrakis(4-hexylphenyl)-dithieno[2,3-d:2 ,3 -d]-s-indaceno[1,2-b:5,6-b]dithiophene; ITIC-2 Cl
Molecular Weight (g/mol) 1565.72
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.