Learn More
Aviva Systems Biology Corp ITIH1 Antibody - C-terminal region (ARP59204_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59204_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITIH1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ITIH1. Alternate names: H1P, IATIH, IGHEP1, ITIH, MGC126415, SHAP, ITI-HC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-616