Learn More
ITL BioMedical Safety SubCulture Unit
Catalog No. 22799900 Shop All ITL BioMedical Products
Change view
Click to view available options
Product Type:
Safety Subculture Unit
Catalog No. Product Type
22-799-900 Safety Subculture Unit
1 options
Catalog No. 22-799-900 Supplier ITL BioMedical Supplier No. A100720
Add to Cart
Add to Cart
Needleless safety device for subculturing blood culture bottles.
Used to transfer sample from positive blood culture bottles for further testing, bringing a new level of safety to microbiology labs.
  • Easy to use and versatile
  • Two procedure options to obtain samples for further testing: drop sample method and syringe method
  • Neither method requires needle which reduces the risk of potential needle stick injury
  • Does not contain latex
  • Compatible fit with various culture bottles
  • Limits exposure to gas or culture discharge
  • Single use only
Specifications
Product Type Safety Subculture Unit