Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITL BioMedical SampLok™ Tube Holders

Catalog No. 21010220 Shop All ITL BioMedical Products
Change view
Click to view available options
Includes:
Integrated safety lid
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Sterility
21-010-220 Integrated safety lid Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-010-220 Supplier ITL BioMedical Supplier No. A100422
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Multi-sample tube holder to collect blood samples into vacuum test tubes and long neck blood culture bottles

  • Ready-to-use
  • Latex free
  • Translucent for full visibility
  • Full length holder reduces potential exposure to sampling needle

A100500

  • Pre-attached luer
  • Sterile, individual package
A100422
  • Integrated safety lid to reduce risk of potential needlestick injury

Specifications

Sterility Non-sterile
Product Type Tube Holder
Color Translucent
For Use With (Application) For collect Blood samples into vacuum test tubes and long neck Blood culture bottles
Latex Free Yes
Includes Integrated safety lid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.