Learn More
Aviva Systems Biology Corp ITLN1 Antibody - middle region (ARP41613_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41613_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITLN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ITLN1. Alternate names: FLJ20022, HL-1, HL1, INTL, ITLN, LFR, hIntL, omentinApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-284-947