Learn More
Aviva Systems Biology Corp ITLN2 Antibody - middle region (ARP50278_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50278_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITLN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ITLN2. Alternate names: HL-2, HL2ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-502