Learn More
Nanocs ITO COATED GLASS SLIDES, 1.1 M
Catalog No. NC0823215
Encompass
Nanocs is a leading provider of Indium Tin Oxide (ITO) coated glass slides for the worldwide researchers use. ITO glass has highly electrical conductivity yet with excellent transparence. It has been widely used in flat panel display and solar cells. The surface resistance of our ITO can be ranged from 8~100 Ohm/sq with the optical transmittance over 85%. Larger sized ITO glass is available upon request. Besides ITO glass, we also provide numerous surface functionalized glass slides including gold coating slides. ITO coated glass slides, 1.1 mm, 25 slides. Resistance 50 (ohm/sq) Size, 1x3"
Catalog No. NC0823215 Supplier Nanocs Supplier No. IT5011125
Nanocs is a leading provider of Indium Tin Oxide (ITO) coated glass slides for the worldwide researchers use. ITO glass has highly electrical conductivity yet with excellent transparence. It has been widely used in flat panel display and solar cells. The surface resistance of our ITO can be ranged from 8~100 Ohm/sq with the optical transmittance over 85%. Larger sized ITO glass is available upon request. Besides ITO glass, we also provide numerous surface functionalized glass slides including gold coating slides. ITO coated glass slides, 1.1 mm, 25 slides. Resistance 50 (ohm/sq) Size, 1x3"