Learn More
Delta Technologies Ltd Polished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated/Indium Tin Oxide coated on polished surface, Rs = 30 - 60 ohms, cut edges. Available quantities: multiples of 10, 25, 100, 250, 1,000+
SDP
Catalog No. NC0719458
Encompass
This item has a minimum qty of 12 per supplier requirements.
ITO Coated Slides, Polished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated/Indium Tin Oxide coated on polished surface, Resistance: 30 - 60 ohms/sq. Delta Technologies, Limited offers a variety of transparent, conductive indium tin oxide (ITO), flourine-doped tin oxide (FTO) coated, and uncoated products on several different substrates, custom cut to any dimension required.
Catalog No. NC0719458 Supplier Delta Technologies Ltd Supplier No. CG81INS115
This item has a minimum qty of 12 per supplier requirements.
ITO Coated Slides, Polished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated/Indium Tin Oxide coated on polished surface, Resistance: 30 - 60 ohms/sq. Delta Technologies, Limited offers a variety of transparent, conductive indium tin oxide (ITO), flourine-doped tin oxide (FTO) coated, and uncoated products on several different substrates, custom cut to any dimension required.