Learn More
Delta Technologies Ltd Polished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated/Indium Tin Oxide coated on polished surface, Rs = 30 - 60 ohms, cut edges. Pack of 100 slides.
SDP
Catalog No. NC1501896
Encompass
ITO Coated Slides, Polished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated/Indium Tin Oxide coated on polished surface, resistance: 30-60 ohms/sq. This is for a box of 100 slides. Delta Technologies, Limited offers a variety of transparent, conductive indium tin oxide (ITO), flourine-doped tin oxide (FTO) coated, and uncoated products on several different substrates, custom cut to any dimension required.
Catalog No. NC1501896 Supplier Delta Technologies Ltd Supplier No. CG81INS115100
ITO Coated Slides, Polished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated/Indium Tin Oxide coated on polished surface, resistance: 30-60 ohms/sq. This is for a box of 100 slides. Delta Technologies, Limited offers a variety of transparent, conductive indium tin oxide (ITO), flourine-doped tin oxide (FTO) coated, and uncoated products on several different substrates, custom cut to any dimension required.