Learn More
Medchemexpress LLC Itopride hydrochloride | 122892-31-3 | 99.8% | 500 MG
SDP
Catalog No. 5000406855
Encompass_Preferred
Itopride (HSR803) hydrochloride is identified as a potent dopamine-2 antagonist and an acetylcholine esterase (AChE) inhibitor. It functions as a gastrointestinal prokinetic agent by enhancing gastric motility through both antidopaminergic and anti-acetylcholinesterasic actions. This compound is also suitable for research related to gastro-esophageal reflux disease (GERD).
  • Potent dopamine-2 antagonist.
  • Acetylcholine esterase (AChE) inhibitor.
  • Enhances gastric motility through antidopaminergic and anti-acetylcholinesterasic actions.
  • Can be utilized as a gastrointestinal prokinetic agent.
  • Suitable for researching gastro-esophageal reflux disease (GERD).
  • Prokinetic effects on both the ileum and colon (in vitro studies).
  • Significantly accelerates gastric emptying (in vivo studies).
Catalog No. 50-004-06855 Supplier Medchemexpress LLC Supplier No. HYB0732500MG
Itopride (HSR803) hydrochloride is identified as a potent dopamine-2 antagonist and an acetylcholine esterase (AChE) inhibitor. It functions as a gastrointestinal prokinetic agent by enhancing gastric motility through both antidopaminergic and anti-acetylcholinesterasic actions. This compound is also suitable for research related to gastro-esophageal reflux disease (GERD).
  • Potent dopamine-2 antagonist.
  • Acetylcholine esterase (AChE) inhibitor.
  • Enhances gastric motility through antidopaminergic and anti-acetylcholinesterasic actions.
  • Can be utilized as a gastrointestinal prokinetic agent.
  • Suitable for researching gastro-esophageal reflux disease (GERD).
  • Prokinetic effects on both the ileum and colon (in vitro studies).
  • Significantly accelerates gastric emptying (in vivo studies).