Learn More
Novus Biologicals™ ITPR2 Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 3709 |
| Species | Human |
| Purity | >50%, by HPLC |
| Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-2466 |
| Gene Alias | inositol 14,5-triphosphate receptor, type 2, inositol 14,5-trisphosphate receptor type 2, insP3R2, IP3 receptor, IP3 receptor isoform 2, IP3R 2, IP3R2, Type 2 inositol 14,5-trisphosphate receptor, Type 2 InsP3 receptor |
| Gene Symbol | ITPR2 |
| Quantity | 0.1 mg |
| Research Category | Neuroscience |
| Storage Buffer | Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.