Learn More
ABclonal Technology ITPR3 Rabbit mAb
SDP
Catalog No. 5000006427
Encompass_Preferred
This gene encodes a receptor for inositol 1 4 5-trisphosphate a second messenger that mediates the release of intracellular calcium The receptor contains a calcium channel at the C-terminus and the ligand-binding site at the N-terminus Knockout studies in mice suggest that type 2 and type 3 inositol 1 4 5-trisphosphate receptors play a key role in exocrine secretion underlying energy metabolism and growth
Catalog No. 50-000-06427 Supplier ABclonal Technology Supplier No. A23202100UL
This gene encodes a receptor for inositol 1 4 5-trisphosphate a second messenger that mediates the release of intracellular calcium The receptor contains a calcium channel at the C-terminus and the ligand-binding site at the N-terminus Knockout studies in mice suggest that type 2 and type 3 inositol 1 4 5-trisphosphate receptors play a key role in exocrine secretion underlying energy metabolism and growth