Learn More
Aviva Systems Biology Corp ITPRIP Antibody - C-terminal region (ARP60894_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60894_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITPRIP. It was validated on Western Blot. . Alternate names: DANGER, KIAA1754, bA127L20, bA127L20.2, RP11-127L20.4ApplicationWB. Species reactivity: Human
Catalog No. 50-287-198