Learn More
TARGETMOL CHEMICALS INC Itraconazole 500MG
SDP
Catalog No. 502880708
Encompass_Preferred
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Itraconazole (R51211) is a triazole antifungal agent that inhibits cytochrome P-450-dependent enzymes necessary for ERGOSTEROL synthesis. Purity 99.68%
Catalog No. 50-288-0708 Supplier TARGETMOL CHEMICALS INC Supplier No. T1011500MG
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Itraconazole (R51211) is a triazole antifungal agent that inhibits cytochrome P-450-dependent enzymes necessary for ERGOSTEROL synthesis. Purity 99.68%