Learn More
Medchemexpress LLC Itraconazole-d3 | 1217512-35-0 | 99.6% | 708.65 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17514S15MG
Encompass_Preferred
Itraconazole-d3 (R51211-d3) is the deuterium labeled Itraconazole, a triazole antifungal agent. It acts as a potent and orally active Hedgehog (Hh) signaling pathway antagonist, inhibiting lanosterol 14α-demethylase. Itraconazole also exhibits anticancer and antiangiogenic effects and is an oxysterol-binding protein (OSBP) inhibitor.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
  • Appearance: solid
  • Color: white to off-white
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-86402