Learn More
Medchemexpress LLC Itraconazole-d3 | 1217512-35-0 | MFCD34676907 | 99.6% | 10 MG
SDP
Catalog No. 5000298534
Encompass_Preferred
Itraconazole-d3 is a deuterium-labeled analog of itraconazole intended for research use as a tracer or internal standard in quantitative analyses. It retains the chemical characteristics of itraconazole and is suitable for analytical development, method validation, and tracer studies in mass spectrometry and NMR workflows.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity (99.6%) suitable for analytical workflows.
  • Compatible with LC-MS, GC-MS, and NMR methods.
  • Suitable for method validation, tracer studies, and stability testing.
  • Small-scale solid quantity designed for analytical use.
Catalog No. 50-002-98534 Supplier Medchemexpress LLC Supplier No. HY17514S110MG
Itraconazole-d3 is a deuterium-labeled analog of itraconazole intended for research use as a tracer or internal standard in quantitative analyses. It retains the chemical characteristics of itraconazole and is suitable for analytical development, method validation, and tracer studies in mass spectrometry and NMR workflows.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity (99.6%) suitable for analytical workflows.
  • Compatible with LC-MS, GC-MS, and NMR methods.
  • Suitable for method validation, tracer studies, and stability testing.
  • Small-scale solid quantity designed for analytical use.