Learn More
Med Vet International Virbac Itrafungol Solution, Antifungal, For Oral Use in Cats, 52mL Liquid
Catalog No. 502828796
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Itrafungol (itraconazole) oral solution is indicated for the treatment of dermatophytosis caused by Microsporum canis in cats. See package insert for complete product details.
Catalog No. 50-282-8796 Supplier Med Vet International Supplier No. RXVITRAFUNGOL
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Itrafungol (itraconazole) oral solution is indicated for the treatment of dermatophytosis caused by Microsporum canis in cats. See package insert for complete product details.