Learn More
BIOGEMS INTERNATIONAL INC ITS (Insulin-Transferrin-Selenium) (100x) 100mL
SDP
Catalog No. 501528631
Encompass
Insulin-Transferrin-Selenium (ITS) is a basal media supplement and can be used to reduce the amount of fetal bovine serum (FBS) needed to culture cells. Insulin promotes the uptake of glucose and amino acids, transferrin is a non-toxic iron carrier, selenium is required for the function of glutathione peroxidase and other antioxidant enzymes. The components of the solution are manufactured animal-free and it is provided as a 100x sterile concentrate.
Catalog No. 50-152-8631 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 00102100
Insulin-Transferrin-Selenium (ITS) is a basal media supplement and can be used to reduce the amount of fetal bovine serum (FBS) needed to culture cells. Insulin promotes the uptake of glucose and amino acids, transferrin is a non-toxic iron carrier, selenium is required for the function of glutathione peroxidase and other antioxidant enzymes. The components of the solution are manufactured animal-free and it is provided as a 100x sterile concentrate.