Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ ITS™ Premix Solution

Catalog No. 092001344 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-200-1344 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-200-1344 Supplier MP Biomedicals™ Supplier No. 092001344
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ITS™ Premix Solution stimulates cell proliferation while decreasing substantially the serum requirements for culture of many cell types.

  • ITS Premix will stimulate cell proliferation while decreasing substantially the serum requirements for culture of many cell types as diverse, for example, as contractile rat heart cells and human colon mucosal epithelial cells.
  • Basal media supplemented with ITS Premix and as little as 2% Fetal Bovine Serum support proliferation of many diploid and heteroploid cell lines at rates equivalent to those obtained with 10% serum.
  • Contains 0.5 mg/mL insulin from bovine pancreas, 0.5 mg/mL human transferrin (substantially iron-free) and 0.5μg/mL sodium selenite.
  • Prepared in Earle's Balanced Salt Solution (EBSS) without phenol red.
  • ITS Premix is sterile filtered through a .2 micron filter.
  • It has been tested and found negative for the presence of bacteria, fungi, and mycoplasma.
  • One milliliter of solution is sufficient to supplement one liter of medium.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Sterility Sterile
For Use With (Application) Mammalian Cell Culture
Quantity 5 mL
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.