Learn More
FUJIFILM Biosciences ITS-X Supplement X100
Catalog No. 5000450363
Encompass_Preferred
This product contains insulin 1 mg/mL transferrin apo 0 55 mg/mL sodium selenite 0 67 mg/L and ethanolamine 0 2 mg/mL It is prepared in EBSS It is a supplement for basal medium to reduce the amount of FBS used in cell culture
Catalog No. 50-004-50363 Supplier FUJIFILM Biosciences Supplier No. 09406761
This product contains insulin 1 mg/mL transferrin apo 0 55 mg/mL sodium selenite 0 67 mg/L and ethanolamine 0 2 mg/mL It is prepared in EBSS It is a supplement for basal medium to reduce the amount of FBS used in cell culture