Learn More
Medchemexpress LLC HY-100508 10mg Medchemexpress, ITSA-1 CAS:200626-61-5 Purity:>98%
SDP
Catalog No. 501931859
Encompass_Preferred
Medchemexpress, HY-100508 10mg ITSA-1 CAS:200626-61-5 ITSA-1 is an activator of histone deacetylase (HDAC) , and counteract trichostatin A (TSA)-induced cell cycle arrest, histone acetylation, and transcriptional activation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1859 Supplier Medchemexpress LLC Supplier No. HY10050810MG
Medchemexpress, HY-100508 10mg ITSA-1 CAS:200626-61-5 ITSA-1 is an activator of histone deacetylase (HDAC) , and counteract trichostatin A (TSA)-induced cell cycle arrest, histone acetylation, and transcriptional activation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.