Learn More
Enzo Life Sciences ITSA-1 (25mg). CAS: 200626-61-5
SDP
Supplier:  Enzo Life Sciences BMLGR3500025
Encompass_Preferred
ITSA-1 is a cell permeable benzotriazole amide that counteracts trichostatin A (BML-GR309) induced cell cycle arrest, histone acetylation and transcriptional activation but not the activity of other HDAC inhibitors. Alternative name: N-(1H-Benzotriazol-1-yl)-2,4-dichlorobenzamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1003