Learn More
Abcam ITSA-1, HDAC activator, 25MG
Catalog No. 502705643
Encompass_Preferred
MW 292.12 Da, Purity >98%. Cell-permeable HDAC activator. Selective Trichostatin A (ab120850) suppressor. Counteracts Trichostatin A-induced cell cycle arrest. Inhibits Trichostatin A-induced tubulin acetylation. Shows no effect on the activity of other HDAC inhibitors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5643 Supplier Abcam Supplier No. AB14453525MG
MW 292.12 Da, Purity >98%. Cell-permeable HDAC activator. Selective Trichostatin A (ab120850) suppressor. Counteracts Trichostatin A-induced cell cycle arrest. Inhibits Trichostatin A-induced tubulin acetylation. Shows no effect on the activity of other HDAC inhibitors.
The product is subject to the following: Abcam Restricted Use Statement