Learn More
Aviva Systems Biology Corp ITSN2 Antibody - C-terminal region (ARP65456_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65456_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ITSN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human ITSN2. Alternate names: KIAA1256, PRO2015, SH3D1B, SH3P18, SWA, SWAPApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-501