Learn More
Enterprise Technology Solutions ITW Permatex Inc Degreaser, Heavy-Duty, Citrus, 5 Gal, Orange
SDP
Catalog No. 5000790539
Encompass_Preferred
Heavy-duty, ready-to-use degreaser effectively removes grease and dirt and works as well as traditional solvents. The professional-strength formula hits hard and fast with a powerful, water-soluble solvent and no harsh fumes or unpleasant odors. Bolstered with fast-acting surfactants, Spray Nine Grez-off Heavy-duty Degreaser digs into grime and pries it away. Use on cleaning engines, machinery, tools, workbenches, garage floors/walls, asphalt, stainless steel, chrome, condenser coils, and exhaust hoods. Degreaser contains no petroleum solvents and is VOC-compliant in all 50 states as well as water-based, nonacidic and nonflammable.
Catalog No. 50-007-90539 Supplier Enterprise Technology Solutions Supplier No. PTX22705
Heavy-duty, ready-to-use degreaser effectively removes grease and dirt and works as well as traditional solvents. The professional-strength formula hits hard and fast with a powerful, water-soluble solvent and no harsh fumes or unpleasant odors. Bolstered with fast-acting surfactants, Spray Nine Grez-off Heavy-duty Degreaser digs into grime and pries it away. Use on cleaning engines, machinery, tools, workbenches, garage floors/walls, asphalt, stainless steel, chrome, condenser coils, and exhaust hoods. Degreaser contains no petroleum solvents and is VOC-compliant in all 50 states as well as water-based, nonacidic and nonflammable.