Learn More
Enterprise Technology Solutions ITW Pro Brands Liquid Deodorizer, 1 Gal, Neutral Scent, 4/CT, White/Green
SDP
Catalog No. 5000831597
Encompass_Preferred
Odor digester contains enzyme-producing bacteria that digest malodors and waste caused by dampness, mildew, mold, food, urine, vomit, fecal and organic matter. Nonstaining formula will not stain carpets, upholstery and water-safe surfaces. Odor digester is nontoxic to animals and humans for safety. Odor digester is perfect for use on carpet, upholstery, concrete, cinder blocks, porous surfaces, grout joints, painted surfaces, garbage cans and more.
Catalog No. 50-008-31597 Supplier Enterprise Technology Solutions Supplier No. ITW33601
Odor digester contains enzyme-producing bacteria that digest malodors and waste caused by dampness, mildew, mold, food, urine, vomit, fecal and organic matter. Nonstaining formula will not stain carpets, upholstery and water-safe surfaces. Odor digester is nontoxic to animals and humans for safety. Odor digester is perfect for use on carpet, upholstery, concrete, cinder blocks, porous surfaces, grout joints, painted surfaces, garbage cans and more.