Learn More
Enterprise Technology Solutions ITW Pro Brands Odor Digester, w/ Enzymes, Nontoxic, 32 oz., White
SDP
Catalog No. 5000831594
Encompass_Preferred
Liquid Alive Odor Digester contains enzyme-producing bacteria that digest malodors and waste. Use in schools, hotels, nursing homes and restaurants. Nontoxic formula does not stain carpets and upholstery,
Catalog No. 50-008-31594 Supplier Enterprise Technology Solutions Supplier No. ITW33632
Liquid Alive Odor Digester contains enzyme-producing bacteria that digest malodors and waste. Use in schools, hotels, nursing homes and restaurants. Nontoxic formula does not stain carpets and upholstery,