Learn More
Enterprise Technology Solutions ITW Pro Brands Stainless Steel Cleaner, Aerosol, 16 oz Net, NTL/Blue/Gray
SDP
Catalog No. 5000831687
Encompass_Preferred
Stainless Steel Cleaner cleans, polishes and preserves the high luster of stainless steel. The oil-based formula leaves behind an invisible protective finish on stainless steel surfaces to help resist re-soiling. Remove spot marks, grease, watermarks, film and fingerprints. Polish the surface with a clean, dry cloth for an even higher gloss. Use the NSF Certified cleaner on aluminum, stainless steel, chrome, copper and brass. Cleaner volume is measured in net weight.
Catalog No. 50-008-31687 Supplier Enterprise Technology Solutions Supplier No. ITW20920
Stainless Steel Cleaner cleans, polishes and preserves the high luster of stainless steel. The oil-based formula leaves behind an invisible protective finish on stainless steel surfaces to help resist re-soiling. Remove spot marks, grease, watermarks, film and fingerprints. Polish the surface with a clean, dry cloth for an even higher gloss. Use the NSF Certified cleaner on aluminum, stainless steel, chrome, copper and brass. Cleaner volume is measured in net weight.