Learn More
TARGETMOL CHEMICALS INC IU1 25MG
SDP
Catalog No. 502579554
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IU1(IC50=4.7 u M) a reversible specific human USP14 proteasome inhibitor can penetrate the cell. purity: 99%
Catalog No. 50-257-9554 Supplier TARGETMOL CHEMICALS INC Supplier No. T610725MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IU1(IC50=4.7 u M) a reversible specific human USP14 proteasome inhibitor can penetrate the cell. purity: 99%