Learn More
Medchemexpress LLC HY-122243 5mg Medchemexpress, IU1-47 CAS:670270-31-2 Purity:>98%
SDP
Catalog No. 501932568
Encompass_Preferred
Medchemexpress, HY-122243 5mg IU1-47 CAS:670270-31-2 IU1-47 is a potent and specific USP14 inhibitor with an IC 50 of 0.6 μM. IU1-47 inhibits IsoT/USP5 with an IC 50 of 20 μM. IU1-47 induces tau elimination in cultured neurons [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2568 Supplier Medchemexpress LLC Supplier No. HY1222435MG
Medchemexpress, HY-122243 5mg IU1-47 CAS:670270-31-2 IU1-47 is a potent and specific USP14 inhibitor with an IC 50 of 0.6 μM. IU1-47 inhibits IsoT/USP5 with an IC 50 of 20 μM. IU1-47 induces tau elimination in cultured neurons [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.