Learn More
Selleck Chemical LLC IU1-47
SDP
Catalog No. 5000002439
Encompass_Preferred
IU1-47 is a potent and specific inhibitor of USP14 with an IC50 of 0 6 M IU1-47 shows 33-fold selectivity for USP14 over USP5 (IsoT)
Catalog No. 50-000-02439 Supplier Selleck Chemical LLC Supplier No. S69435MG
IU1-47 is a potent and specific inhibitor of USP14 with an IC50 of 0 6 M IU1-47 shows 33-fold selectivity for USP14 over USP5 (IsoT)