Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IU1, Tocris Bioscience™

Catalog No. p-4816748
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-881-0 10 mg
40-885-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-881-0 Supplier R&D Systems Supplier No. 4088/10
This item is not returnable. View return policy

USP14 inhibitor

Selective inhibitor of Usp14; inhibits the catalytic activity of proteasome-associated Usp14 in vitro (IC50 < 4 μM). Stimulates proteasome activity and substrate degradation.

Chemical Identifiers

CAS 314245-33-5
Molecular Formula C18H21FN2O
Molecular Weight (g/mol) 300.38
MDL Number MFCD01917473
InChI Key JUWDSDKJBMFLHE-UHFFFAOYSA-N
Synonym iu1, 1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-pyrrolidin-1-yl ethan-1-one, 1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-pyrrolidin-1-yl ethanone, 1-1-4-fluorophenyl-2,5-dimethylpyrrol-3-yl-2-pyrrolidin-1-ylethanone, 1-1-4-fluoro-phenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-pyrrolidin-1-yl-ethanone, 1-1-4-fluorophenyl-2,5-dimethyl-1h-pyrrol-3-yl-2-1-pyrrolidinyl ethanone, usp14 inhibitor, 1-1-4-fluorophenyl-2,5-dimethylpyrrol-3-yl-2-pyrrolidin-1-yl ethanone, iu-1
PubChem CID 675434
IUPAC Name 1-[1-(4-fluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2-(pyrrolidin-1-yl)ethan-1-one
SMILES CC1=CC(C(=O)CN2CCCC2)=C(C)N1C1=CC=C(F)C=C1

Specifications

Quantity 10 mg
Formula Weight 300.37
Percent Purity >98%
Chemical Name or Material IU1
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.