Learn More
Sklar Instruments IUD Extractor, Sklar, 10-1/2 in., Hook Point, Slightly Angled, Horizontally Serrated, Stainless Steel. EA/1.
SDP
Catalog No. 501436860 Shop All Sklar Instruments Products
Encompass_Preferred
IUD Extractor, Sklar, 10-1/2 in., Hook Point, Slightly Angled, Horizontally Serrated, Stainless Steel. Sklar's IUD Extractor is an OB/GYN instrument used for removal of an intrauterine device. It is a narrow instrument with a slight angle in the shaft and two crochet style hooks on each side of the tip used to grasp the IUD strings. This product is straight with a round tip and 10-1/2 inches long.
This item is not returnable. View return policy
Catalog No. 50-143-6860 Supplier Sklar Instruments Supplier No. 908210
IUD Extractor, Sklar, 10-1/2 in., Hook Point, Slightly Angled, Horizontally Serrated, Stainless Steel. Sklar's IUD Extractor is an OB/GYN instrument used for removal of an intrauterine device. It is a narrow instrument with a slight angle in the shaft and two crochet style hooks on each side of the tip used to grasp the IUD strings. This product is straight with a round tip and 10-1/2 inches long.