Learn More
Metrohm USA IUNITRODE WITH PT1000
SDP
Supplier:  Metrohm USA 60278300
Encompass
Combined pH glass electrode for measurements in difficult media, with memory chip for sensor data such as article and serial number, calibration data and calibration history.
Catalog No. NC0186675