Learn More
Med Vet International IV Administration Set, Sterile, 20dr/mL, 72" with Injection Site, Luer Slip, 5 Pack
Catalog No. 503045518
Encompass_Preferred
Our IV administration sets feature universal kink-resistant tubing with smooth action roller clamp, and Y injection site. 20 drops per mL rate. Compatible with both bottles and bags. Roller clamp controls flow rate of intravenous medication or fluids. Sterile Luer Slip Meets a wide range of solution administration requirements. Tubing Length: 72"
Catalog No. 50-304-5518 Supplier Med Vet International Supplier No. IV20X5
Our IV administration sets feature universal kink-resistant tubing with smooth action roller clamp, and Y injection site. 20 drops per mL rate. Compatible with both bottles and bags. Roller clamp controls flow rate of intravenous medication or fluids. Sterile Luer Slip Meets a wide range of solution administration requirements. Tubing Length: 72"