Learn More
Med Vet International IV Administration Set, Sterile, 60dr/mL 72in with Injection Site, Luer Slip, 50/Case
Supplier:  Med Vet International IV60X50
Encompass_Preferred
Our IV administration sets feature universal kink-resistant tubing with smooth action roller clamp, and Y injection site. 60 drops per mL rate. Compatible with both bottles and bags. Roller clamp controls flow rate of intravenous medication or fluids. Sterile Luer Slip Meets a wide range of solution administration requirements. Tubing Length: 72'
Catalog No. 50-264-3570