Learn More
McKesson General Medical IV ADMINISTRATION SET
Supplier:  McKesson General Medical 297692
Encompass_Preferred
Primary IV Administration Set Continu-Flo® Gravity 3 Ports 10 Drops / mL Drip Rate Without Filter 106 Inch Tubing Solution
Catalog No. NC3943682