Learn More
Instech IV bag spike and 30in extension set with pinch clamp ending in male luer. Connects saline bag to sampler tubing on Instech ABS1 and ABS2 automated blood samplers. Sterile.
SDP
Catalog No. 5080982
Encompass
The Instech ABS2/IVEXT is a sterile IV bag spike and 30in extension set designed to connect a saline bag to the sampler tubing on Automated Blood Samplers™ (ABS1™ and ABS2™). Its integrated pinch clamp and male luer fitting provide controlled flow adjustment and a secure connection for reliable IV fluid delivery during automated sampling workflows.
Features
  • Sterile IV bag spike with 30in extension tubing
  • Integrated pinch clamp for flow control
  • Ends in male luer for secure sampler connection
  • Interfaces with ABS1 and ABS2 sampler tubing
  • Supports consistent IV fluid delivery during studies
For animal research only. Not for human use.
Catalog No. 50-809-82 Supplier Instech Supplier No. ABS2/IVEXT
The Instech ABS2/IVEXT is a sterile IV bag spike and 30in extension set designed to connect a saline bag to the sampler tubing on Automated Blood Samplers™ (ABS1™ and ABS2™). Its integrated pinch clamp and male luer fitting provide controlled flow adjustment and a secure connection for reliable IV fluid delivery during automated sampling workflows.
Features
  • Sterile IV bag spike with 30in extension tubing
  • Integrated pinch clamp for flow control
  • Ends in male luer for secure sampler connection
  • Interfaces with ABS1 and ABS2 sampler tubing
  • Supports consistent IV fluid delivery during studies
For animal research only. Not for human use.