Learn More
Med Vet International Nipro Safelet IV Catheter, 20g X 1 in., 50/Box
Catalog No. 502832334
Encompass_Preferred
All Safelet IV catheters are designed specifically to perform in delicate vasculature, delivering maximum efficiency and patient comfort. Smooth catheter-to-needle transition results in atraumatic insertion for optimum patient comfort. ETFE catheter material is biocompatible, minimizing risk of irritation and infection while catheter is indwelling. Catheter is radiopaque and features a flashback window for quick visualization of blood.
Catalog No. 50-283-2334 Supplier Med Vet International Supplier No. CI2025BX
All Safelet IV catheters are designed specifically to perform in delicate vasculature, delivering maximum efficiency and patient comfort. Smooth catheter-to-needle transition results in atraumatic insertion for optimum patient comfort. ETFE catheter material is biocompatible, minimizing risk of irritation and infection while catheter is indwelling. Catheter is radiopaque and features a flashback window for quick visualization of blood.