Learn More
Med Vet International Nipro IV Catheters, 24 X 1", 50/Box
Catalog No. 502832112
Encompass_Preferred
All Safelet IV catheters are designed specifically to perform in delicate vasculature, delivering maximum efficiency and patient comfort. Smooth catheter-to-needle transition results in atraumatic insertion for optimum patient comfort. ETFE catheter material is biocompatible, minimizing risk of irritation and infection while catheter is indwelling. Catheter is radiopaque and features a flashback window for quick visualization of blood. Hub is color-coded for rapid identification of catheter size. Available in a wide selection of sizes to match virtually any clinical need.
Catalog No. 50-283-2112 Supplier Med Vet International Supplier No. CI2425BX
All Safelet IV catheters are designed specifically to perform in delicate vasculature, delivering maximum efficiency and patient comfort. Smooth catheter-to-needle transition results in atraumatic insertion for optimum patient comfort. ETFE catheter material is biocompatible, minimizing risk of irritation and infection while catheter is indwelling. Catheter is radiopaque and features a flashback window for quick visualization of blood. Hub is color-coded for rapid identification of catheter size. Available in a wide selection of sizes to match virtually any clinical need.