Learn More
Med Vet International Terumo Surflo IV Catheter, 24G x .75in
Catalog No. 502092366
Encompass_Preferred
The Terumo Surflo is a .75in peripheral IV equipped with an ultra-sharp double-bevel 24-gauge introducer needle coated with a medical-grade lubricant to help prevent inflammation as much as possible. Used to administer medicine intravenously. This is a flexible catheter with a large inner-wall diameter assuring good, stable blood flow. A preview chamber in clear sure-grip gives immediate indication of proper placement. Low risk of split tips and peel back. Comes individually in easy to handle, tamper-evident packaging.
This item is not returnable. View return policy
Catalog No. 50-209-2366 Supplier Med Vet International Supplier No. SROX2419CA
The Terumo Surflo is a .75in peripheral IV equipped with an ultra-sharp double-bevel 24-gauge introducer needle coated with a medical-grade lubricant to help prevent inflammation as much as possible. Used to administer medicine intravenously. This is a flexible catheter with a large inner-wall diameter assuring good, stable blood flow. A preview chamber in clear sure-grip gives immediate indication of proper placement. Low risk of split tips and peel back. Comes individually in easy to handle, tamper-evident packaging.