Learn More
Health Care Logistics Inc IV COVERS, AMBER, 5 X 8-1/2
SDP
Supplier:  Health Care Logistics Inc 7586
Encompass
UV Blocking - Covers block 90% of UV light and are perfect for dispensing IV admixtures that contain light-sensitive drugs.Opacity - IV Covers are Light Amber with slit at sealed endTransparent - Bags are transparent, but will mask many differences in IV fluid color.Thickness - 1.5-mil - 1.8 milQuantity - 100 per packDimensions: 5"W x 8½"H
Catalog No. NC1959954