Learn More
Health Care Logistics Inc IV Covers, Green, 6 x 10, Slit at Sealed End | 100/pack
SDP
Supplier:  Health Care Logistics Inc 7584
Encompass
UV Blocking - Covers block 90% of UV light and are perfect for dispensing IV admixtures that contain light-sensitive drugs. | Opacity - IV Covers are Green with slit at sealed end | Transparent - Bags are transparent, but will mask many differences in IV fluid color. | Thickness - 1.5-mil - 1.8 mil | Dimensions: 6"W x 10"H
Catalog No. NC1775212