Learn More
SAI Infusion Technologies Micro Flow Extension Line - 45", Male & Female Luers
SDP
Catalog No. NC0628427
Encompass
45" Micro Flow Extension line with male and female luer adapters, 225ul dead volume. SAI extension lines are ideal for connecting between pump and swivel or pump and catheter in animal infusion studies. Made with non-DEHP PVC tubing and available in a variety of standard lengths. Custom sizes are also available. 1/each. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC0628427 Supplier SAI Infusion Technologies Supplier No. EXT45G
45" Micro Flow Extension line with male and female luer adapters, 225ul dead volume. SAI extension lines are ideal for connecting between pump and swivel or pump and catheter in animal infusion studies. Made with non-DEHP PVC tubing and available in a variety of standard lengths. Custom sizes are also available. 1/each. Product is for laboratory animal research only.