Learn More
Bound Tree Medical IV EXTENSION SET LL M 8IN
Catalog No. NC2108808
Encompass_Preferred
Curaplex® IV Extension Set with Removable Sure-Lock, Needle-Free Connector, Pinch Clamp, Rotating Male Luer-Lock, 8in
This item is not returnable. View return policy
Catalog No. NC2108808 Supplier Bound Tree Medical Supplier No. 171431081
Curaplex® IV Extension Set with Removable Sure-Lock, Needle-Free Connector, Pinch Clamp, Rotating Male Luer-Lock, 8in