Learn More
Med Vet International Standard Bore "T"-Port Extension, J0467F
Catalog No. 502829759
Encompass_Preferred
Ultrasite female luer lock needle free. Spin male luer lock. Latex injection port. 7.5" length (19.1cm). 1.3ml priming volume. Slide clamp.
Catalog No. 50-282-9759 Supplier Med Vet International Supplier No. JOR467F
Add to Cart
Add to Cart
Ultrasite female luer lock needle free. Spin male luer lock. Latex injection port. 7.5" length (19.1cm). 1.3ml priming volume. Slide clamp.