Learn More
Med Vet International JorVet IV Extension Set w/ Flow Control, J1038
Supplier:  Med Vet International JOR1038
Encompass_Preferred
With flow rate regulator, this 19" extension set has an adjustable dial that controls the flow rate in mL/min. It has a female luer fitting on one end and a male spin luer lock on the other. A "Y" site is located just above below the controller.
Catalog No. NC2290918