Learn More
McKesson General Medical IV PMP SET ALARIS PUMP 3 PORTS
Catalog No. NC3522171
Encompass
IV Pump Set Alaris® Pump 3 Ports 20 Drops / mL Drip Rate 0.2 Micron Filter 117 Inch Tubing Solution
Catalog No. NC3522171 Supplier McKesson General Medical Supplier No. 832456
IV Pump Set Alaris® Pump 3 Ports 20 Drops / mL Drip Rate 0.2 Micron Filter 117 Inch Tubing Solution