Learn More
Med Vet International IV Pump, J0027R
Catalog No. 5000585454
Encompass_Preferred
For rapid infusion of large volumes of IV fluids, the rubber bulb pump infuses air into any bottle by needle insertion. Air pressure forces fluid out much quicker than simple gravity. For equine and bovine use. Dark orange.
Catalog No. 50-005-85454 Supplier Med Vet International Supplier No. JOR27R
For rapid infusion of large volumes of IV fluids, the rubber bulb pump infuses air into any bottle by needle insertion. Air pressure forces fluid out much quicker than simple gravity. For equine and bovine use. Dark orange.